missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AAA1928006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1928006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1928006 Supplier Thermo Scientific Chemicals Supplier No. A1928006
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 202865-68-7
Molecular Formula C7H8BrClFN
Molecular Weight (g/mol) 240.50
MDL Number MFCD00143094
InChI Key RSFNYNQGERWCMT-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorobenzylamine hydrochloride, 3-bromo-4-fluorophenyl methanamine hydrochloride, 3-bromo-4-fluorobenzylamine hcl, 1-3-bromo-4-fluorophenyl methanamine hydrochloride, 3-bromo-4-fluorophenyl methylamine, chloride, 1-3-bromo-4-fluorophenyl methylamine hydrochloride, benzenemethanamine, 3-bromo-4-fluoro-, hydrochloride, pubchem3226, 3-bromo-4-fluorobenzylaminehydrochloride, acmc-1bczm
PubChem CID 2724901
IUPAC Name (3-bromo-4-fluorophenyl)methanamine;hydrochloride
SMILES Cl.NCC1=CC(Br)=C(F)C=C1

Specifications

Melting Point 214°C to 218°C
Quantity 5 g
Formula Weight 240.5
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-fluorobenzylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only