missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzyl bromide, 97%

CAS: 78239-71-1 | C7H5Br2F | 267.92 g/mol

$61.62 - $603.74

Chemical Identifiers

CAS 78239-71-1
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.92
MDL Number MFCD09037367
InChI Key ZRWSODQPUJMFRV-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorobenzyl bromide, 2-bromo-4-bromomethyl-1-fluorobenzene, 3-bromo-4-fluorobenzylbromide, benzene, 2-bromo-4-bromomethyl-1-fluoro, 2-bromanyl-4-bromomethyl-1-fluoranyl-benzene, pubchem24015, acmc-209pdt, 3-bromo-4-fluoro-benzyl bromide, 2-bromo-4-bromomethyl-1-fluoro-benzene, benzene,2-bromo-4-bromomethyl-1-fluoro
PubChem CID 12922710
IUPAC Name 2-bromo-4-(bromomethyl)-1-fluorobenzene
SMILES FC1=C(Br)C=C(CBr)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6139906
View Documents
Thermo Scientific Chemicals
H6139906
5 g
Each for $61.62
Only null left
 
AAH6139914
View Documents
Thermo Scientific Chemicals
H6139914
25 g
Each for $211.84
Only null left
 
AAH6139922
View Documents
Thermo Scientific Chemicals
H6139922
100 g
Each for $603.74
Only null left
 
Description

Description

3-Bromo-4-fluorobenzyl bromide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-fluorobenzyl bromide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive. Moisture sensitive. Store under Argon.
Specifications

Chemical Identifiers

78239-71-1
267.92
ZRWSODQPUJMFRV-UHFFFAOYSA-N
12922710
FC1=C(Br)C=C(CBr)C=C1
C7H5Br2F
MFCD09037367
3-bromo-4-fluorobenzyl bromide, 2-bromo-4-bromomethyl-1-fluorobenzene, 3-bromo-4-fluorobenzylbromide, benzene, 2-bromo-4-bromomethyl-1-fluoro, 2-bromanyl-4-bromomethyl-1-fluoranyl-benzene, pubchem24015, acmc-209pdt, 3-bromo-4-fluoro-benzyl bromide, 2-bromo-4-bromomethyl-1-fluoro-benzene, benzene,2-bromo-4-bromomethyl-1-fluoro
2-bromo-4-(bromomethyl)-1-fluorobenzene

Specifications

78239-71-1
White
MFCD09037367
UN3261
Slightly soluble in water.
FC1=C(Br)C=C(CBr)C=C1
267.92
267.92
Solid
41°C
C7H5Br2F
5 g
3-bromo-4-fluorobenzyl bromide, 2-bromo-4-bromomethyl-1-fluorobenzene, 3-bromo-4-fluorobenzylbromide, benzene, 2-bromo-4-bromomethyl-1-fluoro, 2-bromanyl-4-bromomethyl-1-fluoranyl-benzene, pubchem24015, acmc-209pdt, 3-bromo-4-fluoro-benzyl bromide, 2-bromo-4-bromomethyl-1-fluoro-benzene, benzene,2-bromo-4-bromomethyl-1-fluoro
ZRWSODQPUJMFRV-UHFFFAOYSA-N
2-bromo-4-(bromomethyl)-1-fluorobenzene
12922710
97%
3-Bromo-4-fluorobenzyl bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only