missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzyl bromide, 97%

CAS: 78239-71-1 | C7H5Br2F | 267.92 g/mol

Supplier:  Thermo Scientific Chemicals H6139914

 View more versions of this product

Catalog No. AAH6139914


Only null left
Explore available promotions

Description

Description

3-Bromo-4-fluorobenzyl bromide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-fluorobenzyl bromide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive. Moisture sensitive. Store under Argon.
Specifications

Specifications

3-Bromo-4-fluorobenzyl bromide
41°C
C7H5Br2F
25 g
3-bromo-4-fluorobenzyl bromide, 2-bromo-4-bromomethyl-1-fluorobenzene, 3-bromo-4-fluorobenzylbromide, benzene, 2-bromo-4-bromomethyl-1-fluoro, 2-bromanyl-4-bromomethyl-1-fluoranyl-benzene, pubchem24015, acmc-209pdt, 3-bromo-4-fluoro-benzyl bromide, 2-bromo-4-bromomethyl-1-fluoro-benzene, benzene,2-bromo-4-bromomethyl-1-fluoro
ZRWSODQPUJMFRV-UHFFFAOYSA-N
2-bromo-4-(bromomethyl)-1-fluorobenzene
12922710
97%
78239-71-1
White
MFCD09037367
UN3261
Slightly soluble in water.
FC1=C(Br)C=C(CBr)C=C1
267.92
267.92
Solid

Chemical Identifiers

78239-71-1
267.92
ZRWSODQPUJMFRV-UHFFFAOYSA-N
12922710
FC1=C(Br)C=C(CBr)C=C1
C7H5Br2F
MFCD09037367
3-bromo-4-fluorobenzyl bromide, 2-bromo-4-bromomethyl-1-fluorobenzene, 3-bromo-4-fluorobenzylbromide, benzene, 2-bromo-4-bromomethyl-1-fluoro, 2-bromanyl-4-bromomethyl-1-fluoranyl-benzene, pubchem24015, acmc-209pdt, 3-bromo-4-fluoro-benzyl bromide, 2-bromo-4-bromomethyl-1-fluoro-benzene, benzene,2-bromo-4-bromomethyl-1-fluoro
2-bromo-4-(bromomethyl)-1-fluorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only