missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzonitrile 99.0+%, TCI America™

Catalog No. B196525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B196525G 25 g
B19655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B196525G Supplier TCI America Supplier No. B196525G

Chemical Identifiers

CAS 79630-23-2
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00055432
InChI Key JKCYKISVUIVZCS-UHFFFAOYSA-N
Synonym 5-cyano-2-fluorobromobenzene, benzonitrile, 3-bromo-4-fluoro, 3-bromo-4-fluorobenzenecarbonitrile, 4-fluoro-3-bromobenzonitrile, 3-bromo-4-fluoro-benzonitrile, pubchem1580, 3-bromo-4-flurobenzonitrile, acmc-1bh34, 3-bromo,4-fluorobenzonitrile, ksc494e4j
PubChem CID 123579
IUPAC Name 3-bromo-4-fluorobenzonitrile
SMILES C1=CC(=C(C=C1C#N)Br)F

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 200.01
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-fluorobenzonitrile