missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzonitrile, 98%

Catalog No. aaa1331806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1331806 5 g
AAA1331814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1331806 Supplier Thermo Scientific Chemicals Supplier No. A1331806
Only null left

CAS: 79630-23-2 | C7H3BrFN | 200.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79630-23-2
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00055432
InChI Key JKCYKISVUIVZCS-UHFFFAOYSA-N
Synonym 5-cyano-2-fluorobromobenzene, benzonitrile, 3-bromo-4-fluoro, 3-bromo-4-fluorobenzenecarbonitrile, 4-fluoro-3-bromobenzonitrile, 3-bromo-4-fluoro-benzonitrile, pubchem1580, 3-bromo-4-flurobenzonitrile, acmc-1bh34, 3-bromo,4-fluorobenzonitrile, ksc494e4j
PubChem CID 123579
IUPAC Name 3-bromo-4-fluorobenzonitrile
SMILES C1=CC(=C(C=C1C#N)Br)F

Specifications

Melting Point 54°C to 58°C
Boiling Point 134°C to 136°C (33 mmHg)
Quantity 5 g
UN Number UN3439
Beilstein 8198509
Formula Weight 200.01
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-fluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 279-200-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only