missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-chloroanisole, 98%

Catalog No. AAA1866003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1866003 1 g
AAA1866006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1866003 Supplier Thermo Scientific Chemicals Supplier No. A1866003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2732-80-1 | C7H6BrClO | 221.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2732-80-1
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.48
MDL Number MFCD00070739
InChI Key SQHMXVXKKCXIGN-UHFFFAOYSA-N
Synonym 3-bromo-4-chloroanisole, 4-chloro-3-bromoanisole, 2-chloro-5-methoxybromobenzene, benzene, 2-bromo-1-chloro-4-methoxy, 3-bromo-4-chlorophenyl methyl ether, pubchem4555, acmc-1clwv, 3-bromo-4-chloroamizole, ksc497k1d, 2-chloro-5-methoxy-bromobenzene
PubChem CID 24212081
IUPAC Name 2-bromo-1-chloro-4-methoxybenzene
SMILES COC1=CC(Br)=C(Cl)C=C1

Specifications

Quantity 1 g
Formula Weight 221.48
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-chloroanisole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only