missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-fluorotoluene 98.0+%, TCI America™
$53.90 - $161.72
Chemical Identifiers
| CAS | 59907-12-9 |
|---|---|
| Molecular Formula | C7H6BrF |
| Molecular Weight (g/mol) | 189.027 |
| MDL Number | MFCD04972694 |
| InChI Key | LZVNGSFAHGKCDM-UHFFFAOYSA-N |
| Synonym | 3-bromo-2-fluorotoluene, 1-bromo-2-fluoro-3-methyl-benzene, benzene, 1-bromo-2-fluoro-3-methyl, 2-fluoro-3-bromo toluene, 2-fluoro-3-methyl-bromobenzene, pubchem4521, a-bromo-a-fluorotoluene, 2-fluoro-3-bromotoluene, acmc-1b1zk, ksc493o4d |
| PubChem CID | 108855 |
| IUPAC Name | 1-bromo-2-fluoro-3-methylbenzene |
| SMILES | CC1=C(C(=CC=C1)Br)F |
Chemical Identifiers
| 59907-12-9 | |
| 189.027 | |
| LZVNGSFAHGKCDM-UHFFFAOYSA-N | |
| 108855 | |
| CC1=C(C(=CC=C1)Br)F |
| C7H6BrF | |
| MFCD04972694 | |
| 3-bromo-2-fluorotoluene, 1-bromo-2-fluoro-3-methyl-benzene, benzene, 1-bromo-2-fluoro-3-methyl, 2-fluoro-3-bromo toluene, 2-fluoro-3-methyl-bromobenzene, pubchem4521, a-bromo-a-fluorotoluene, 2-fluoro-3-bromotoluene, acmc-1b1zk, ksc493o4d | |
| 1-bromo-2-fluoro-3-methylbenzene |
Specifications
| 59907-12-9 | |
| 186°C | |
| MFCD04972694 | |
| 3-bromo-2-fluorotoluene, 1-bromo-2-fluoro-3-methyl-benzene, benzene, 1-bromo-2-fluoro-3-methyl, 2-fluoro-3-bromo toluene, 2-fluoro-3-methyl-bromobenzene, pubchem4521, a-bromo-a-fluorotoluene, 2-fluoro-3-bromotoluene, acmc-1b1zk, ksc493o4d | |
| CC1=C(C(=CC=C1)Br)F | |
| 189.027 | |
| 189.03 | |
| Liquid |
| Yellow | |
| C7H6BrF | |
| 1 g | |
| LZVNGSFAHGKCDM-UHFFFAOYSA-N | |
| 1-bromo-2-fluoro-3-methylbenzene | |
| 108855 | |
| ≥98.0% (GC) | |
| 3-Bromo-2-fluorotoluene |
Safety and Handling
TSCA : No