missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-chloro-5-methylpyridine 96.0+%, TCI America™
Supplier: TCI America B506125G
Specifications
| 3-Bromo-2-chloro-5-methylpyridine | |
| 67°C | |
| MFCD01830664 | |
| 2-chloro-3-bromo-5-methylpyridine, 3-bromo-2-chloro-5-picoline, 5-bromo-6-chloro-3-picoline, pyridine, 3-bromo-2-chloro-5-methyl, 3-bromo-2-chloro-5-methyl-pyridine, 2-chloro-5-methyl-3-bromopyridine, pubchem1167, acmc-1cbw1, ksc490s4b, 2-chloro-3-bromo-5-picoline | |
| CC1=CC(=C(N=C1)Cl)Br | |
| 206.467 | |
| 206.47 | |
| Crystalline Powder |
| 17282-03-0 | |
| C6H5BrClN | |
| 25 g | |
| ISNRJFOYJNIKTN-UHFFFAOYSA-N | |
| 3-bromo-2-chloro-5-methylpyridine | |
| 285437 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 17282-03-0 | |
| 206.467 | |
| ISNRJFOYJNIKTN-UHFFFAOYSA-N | |
| 285437 | |
| CC1=CC(=C(N=C1)Cl)Br |
| C6H5BrClN | |
| MFCD01830664 | |
| 2-chloro-3-bromo-5-methylpyridine, 3-bromo-2-chloro-5-picoline, 5-bromo-6-chloro-3-picoline, pyridine, 3-bromo-2-chloro-5-methyl, 3-bromo-2-chloro-5-methyl-pyridine, 2-chloro-5-methyl-3-bromopyridine, pubchem1167, acmc-1cbw1, ksc490s4b, 2-chloro-3-bromo-5-picoline | |
| 3-bromo-2-chloro-5-methylpyridine |
Safety and Handling
TSCA : No