missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-chloro-5-methylpyridine 96.0+%, TCI America™

Catalog No. p-7123928
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50615G 5 g
B506125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50615G Supplier TCI America Supplier No. B50615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17282-03-0 | C6H5BrClN | 206.467 g/mol

Chemical Identifiers

CAS 17282-03-0
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD01830664
InChI Key ISNRJFOYJNIKTN-UHFFFAOYSA-N
Synonym 2-chloro-3-bromo-5-methylpyridine, 3-bromo-2-chloro-5-picoline, 5-bromo-6-chloro-3-picoline, pyridine, 3-bromo-2-chloro-5-methyl, 3-bromo-2-chloro-5-methyl-pyridine, 2-chloro-5-methyl-3-bromopyridine, pubchem1167, acmc-1cbw1, ksc490s4b, 2-chloro-3-bromo-5-picoline
PubChem CID 285437
IUPAC Name 3-bromo-2-chloro-5-methylpyridine
SMILES CC1=CC(=C(N=C1)Cl)Br

Specifications

Melting Point 67°C
Quantity 5 g
Formula Weight 206.47
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-chloro-5-methylpyridine