accessibility menu, dialog, popup

UserName

CAS RN 17282-03-0

CH3NClBr

CAS RN 17282-03-0

IUPAC Name: 3-bromo-2-chloro-5-methylpyridine
Synonyms: 3-bromo-2-chloro-5-methylpyridine
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: ISNRJFOYJNIKTN-UHFFFAOYSA-N
SMILES: CC1=CN=C(Cl)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results