missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,6-dimethoxytoluene 97.0+%, TCI America™

Catalog No. B4745200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4745200MG 200 mg
B47451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4745200MG Supplier TCI America Supplier No. B4745200MG
Only null left

Chemical Identifiers

CAS 22794-95-2
Molecular Formula C9H11BrO2
Molecular Weight (g/mol) 231.089
InChI Key WXHZGDILTIFQDK-UHFFFAOYSA-N
Synonym 1-Bromo-2,4-dimethoxy-3-methylbenzene
PubChem CID 10889743
IUPAC Name 1-bromo-2,4-dimethoxy-3-methylbenzene
SMILES CC1=C(C=CC(=C1OC)Br)OC

Specifications

Boiling Point 96°C
Quantity 200 mg
Formula Weight 231.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2,6-dimethoxytoluene