accessibility menu, dialog, popup

UserName

CAS RN 22794-95-2

CH3OH3CBrOCH3

CAS RN 22794-95-2

IUPAC Name: 1-bromo-2,4-dimethoxy-3-methylbenzene
Synonyms: 1-bromo-2,4-dimethoxy-3-methylbenzene
Molecular Weight (g/mol): 231.09
Molecular Formula: C9H11BrO2
InChi Key: WXHZGDILTIFQDK-UHFFFAOYSA-N
SMILES: COC1=C(C)C(OC)=C(Br)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results