missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,6-difluorobenzaldehyde, 98%

Catalog No. p-7028805
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAH2621306 5 g
AAH2621314 25 g
2 options

Catalog No. AAH2621306

Supplier: Thermo Scientific Chemicals H2621306

Only null left

CAS: 398456-82-1 | C7H3BrF2O | 221.001 g/mol

3-Bromo-2,6-difluorobenzaldehyde is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2,6-difluorobenzaldehyde is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.

Chemical Identifiers

CAS 398456-82-1
Molecular Formula C7H3BrF2O
Molecular Weight (g/mol) 221.001
MDL Number MFCD08282773
InChI Key OXBHKEYDKAWFLS-UHFFFAOYSA-N
PubChem CID 22139701
IUPAC Name 3-bromo-2,6-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1F)C=O)F)Br

Specifications

Melting Point 53°C to 57°C
Quantity 5 g
Sensitivity Air Sensitive
Synonym 3-bromo-2,6-difluoro-benzaldehyde, benzaldehyde, 3-bromo-2,6-difluoro, acmc-209j8a, 3-bromo-2, 6-difluorobenzaldehyde, benzaldehyde,3-bromo-2,6-difluoro
Formula Weight 221
Percent Purity 98%
Chemical Name or Material 3-Bromo-2,6-difluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only