accessibility menu, dialog, popup

UserName

FOFBr

CAS RN 398456-82-1

IUPAC Name: 3-bromo-2,6-difluorobenzaldehyde
Synonyms: 3-bromo-2,6-difluorobenzaldehyde
Molecular Weight (g/mol): 221
Molecular Formula: C7H3BrF2O
InChi Key: OXBHKEYDKAWFLS-UHFFFAOYSA-N
SMILES: FC1=C(C=O)C(F)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results