missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,6-difluoroaniline, 96%

CAS: 1262198-07-1 | C6H4BrF2N | 208.01 g/mol

$356.59 - $1408.71

Chemical Identifiers

CAS 1262198-07-1
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.01
MDL Number MFCD22627833
InChI Key HIZADWSSPWORHH-UHFFFAOYSA-N
Synonym benzenamine,3-bromo-2,6-difluoro
PubChem CID 21645692
IUPAC Name 3-bromo-2,6-difluoroaniline
SMILES NC1=C(F)C=CC(Br)=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6260903
View Documents
Thermo Scientific Chemicals
H6260903
1 g
Each for $356.59
Only null left
 
AAH6260906
View Documents
Thermo Scientific Chemicals
H6260906
5 g
Each for $1,408.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1262198-07-1
208.01
HIZADWSSPWORHH-UHFFFAOYSA-N
21645692
NC1=C(F)C=CC(Br)=C1F
C6H4BrF2N
MFCD22627833
benzenamine,3-bromo-2,6-difluoro
3-bromo-2,6-difluoroaniline

Specifications

1262198-07-1
MFCD22627833
UN2811
HIZADWSSPWORHH-UHFFFAOYSA-N
3-bromo-2,6-difluoroaniline
21645692
96%
C6H4BrF2N
1 g
benzenamine,3-bromo-2,6-difluoro
NC1=C(F)C=CC(Br)=C1F
208.01
208
3-Bromo-2,6-difluoroaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only