missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,6-difluoroaniline, 96%

CAS: 1262198-07-1 | C6H4BrF2N | 208.01 g/mol

Supplier:  Thermo Scientific Chemicals H6260906

 View more versions of this product

Catalog No. AAH6260906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

3-Bromo-2,6-difluoroaniline
C6H4BrF2N
5 g
benzenamine,3-bromo-2,6-difluoro
NC1=C(F)C=CC(Br)=C1F
208.01
208
1262198-07-1
MFCD22627833
UN2811
HIZADWSSPWORHH-UHFFFAOYSA-N
3-bromo-2,6-difluoroaniline
21645692
96%

Chemical Identifiers

1262198-07-1
208.01
HIZADWSSPWORHH-UHFFFAOYSA-N
21645692
NC1=C(F)C=CC(Br)=C1F
C6H4BrF2N
MFCD22627833
benzenamine,3-bromo-2,6-difluoro
3-bromo-2,6-difluoroaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only