missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2,4,6-trimethylaniline 98.0+%, TCI America™
Supplier: TCI America B384925G
Specifications
| 3-Bromo-2,4,6-trimethylaniline | |
| 38°C | |
| C9H12BrN | |
| 25 g | |
| MVLMPTBHZPYDBZ-UHFFFAOYSA-N | |
| 3-bromo-2,4,6-trimethylaniline | |
| 688300 | |
| ≥98.0% (GC,T) |
| 82842-52-2 | |
| 173°C | |
| MFCD00015476 | |
| benzenamine, 3-bromo-2,4,6-trimethyl, pubchem4363, ksc496e8h, 2,4,6-trimethyl-3-bromoaniline, 3-bromo-2,4,6-trimethyl-aniline, 3-bromo-2,4,6-trimethylbenzenamine, 3-bromo-2,4,6-trimethylphenylamine, 3-bromo-2,4,6-trimethyl-phenylamine, 3-bromo-2,4,6-trimethylaniline, 3-bromo-2,4,6-trimethylaniline, 98+% | |
| CC1=CC(=C(C(=C1N)C)Br)C | |
| 214.106 | |
| 214.11 | |
| Crystalline Powder |
Chemical Identifiers
| 82842-52-2 | |
| 214.106 | |
| MVLMPTBHZPYDBZ-UHFFFAOYSA-N | |
| 688300 | |
| CC1=CC(=C(C(=C1N)C)Br)C |
| C9H12BrN | |
| MFCD00015476 | |
| benzenamine, 3-bromo-2,4,6-trimethyl, pubchem4363, ksc496e8h, 2,4,6-trimethyl-3-bromoaniline, 3-bromo-2,4,6-trimethyl-aniline, 3-bromo-2,4,6-trimethylbenzenamine, 3-bromo-2,4,6-trimethylphenylamine, 3-bromo-2,4,6-trimethyl-phenylamine, 3-bromo-2,4,6-trimethylaniline, 3-bromo-2,4,6-trimethylaniline, 98+% | |
| 3-bromo-2,4,6-trimethylaniline |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator