missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Boc-amino)azetidine hydrochloride, 97%, Thermo Scientific™
$235.03
Chemical Identifiers
| CAS | 217806-26-3 |
|---|---|
| Molecular Formula | C8H17ClN2O2 |
| Molecular Weight (g/mol) | 208.686 |
| MDL Number | MFCD09839423 |
| InChI Key | VLOLOLUAVFGUTD-UHFFFAOYSA-N |
| Synonym | 3-boc-aminoazetidine hydrochloride, tert-butyl azetidin-3-ylcarbamate hydrochloride, 3-n-boc-aminoazetidine hcl, tert-butyl azetidin-3-yl carbamate hydrochloride, tert-butyl n-azetidin-3-yl carbamate hydrochloride, 3-boc-amino azetidine hydrochloride, 3-n-boc-aminoazetidine hydrochloride, 3-boc-amino-azetidine hcl, tert-butyl 3-azetidinylcarbamate hydrochloride, 3-n-boc-amino-azetidine-hcl |
| PubChem CID | 22101213 |
| IUPAC Name | tert-butyl N-(azetidin-3-yl)carbamate;hydrochloride |
| SMILES | CC(C)(C)OC(=O)NC1CNC1.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH59695ME
|
N/A | 500 mg |
Each for $235.03
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 217806-26-3 | |
| 208.686 | |
| VLOLOLUAVFGUTD-UHFFFAOYSA-N | |
| 22101213 | |
| CC(C)(C)OC(=O)NC1CNC1.Cl |
| C8H17ClN2O2 | |
| MFCD09839423 | |
| 3-boc-aminoazetidine hydrochloride, tert-butyl azetidin-3-ylcarbamate hydrochloride, 3-n-boc-aminoazetidine hcl, tert-butyl azetidin-3-yl carbamate hydrochloride, tert-butyl n-azetidin-3-yl carbamate hydrochloride, 3-boc-amino azetidine hydrochloride, 3-n-boc-aminoazetidine hydrochloride, 3-boc-amino-azetidine hcl, tert-butyl 3-azetidinylcarbamate hydrochloride, 3-n-boc-amino-azetidine-hcl | |
| tert-butyl N-(azetidin-3-yl)carbamate;hydrochloride |
Specifications
| 217806-26-3 | |
| C8H17ClN2O2 | |
| 3-boc-aminoazetidine hydrochloride, tert-butyl azetidin-3-ylcarbamate hydrochloride, 3-n-boc-aminoazetidine hcl, tert-butyl azetidin-3-yl carbamate hydrochloride, tert-butyl n-azetidin-3-yl carbamate hydrochloride, 3-boc-amino azetidine hydrochloride, 3-n-boc-aminoazetidine hydrochloride, 3-boc-amino-azetidine hcl, tert-butyl 3-azetidinylcarbamate hydrochloride, 3-n-boc-amino-azetidine-hcl | |
| CC(C)(C)OC(=O)NC1CNC1.Cl | |
| 208.686 | |
| 22101213 | |
| 97% |
| 303°C | |
| MFCD09839423 | |
| VLOLOLUAVFGUTD-UHFFFAOYSA-N | |
| tert-butyl N-(azetidin-3-yl)carbamate;hydrochloride | |
| 137°C (279°F) | |
| 208.69 | |
| 3-(Boc-amino)azetidine hydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures