CAS RN 217806-26-3
IUPAC Name:
tert-butyl N-(azetidin-3-yl)carbamate hydrochloride
Synonyms:
tert-butyl N-(azetidin-3-yl)carbamate hydrochloride
Molecular Weight (g/mol):
208.69
Molecular Formula:
C8H17ClN2O2
InChi Key:
VLOLOLUAVFGUTD-UHFFFAOYSA-N
SMILES:
Cl.CC(C)(C)OC(=O)NC1CNC1
1
–
1
of
1
results
1
Promotions Available
3-(Boc-amino)azetidine hydrochloride, 97%, Thermo Scientific™
3-(Boc-amino)azetidine hydrochloride, 97%, Thermo Scientific™
CAS: 217806-26-3 Molecular Formula: C8H17ClN2O2 Molecular Weight (g/mol): 208.686 MDL Number: MFCD09839423 InChI Key: VLOLOLUAVFGUTD-UHFFFAOYSA-N Synonym: 3-boc-aminoazetidine hydrochloride,tert-butyl azetidin-3-ylcarbamate hydrochloride,3-n-boc-aminoazetidine hcl,tert-butyl azetidin-3-yl carbamate hydrochloride,tert-butyl n-azetidin-3-yl carbamate hydrochloride,3-boc-amino azetidine hydrochloride,3-n-boc-aminoazetidine hydrochloride,3-boc-amino-azetidine hcl,tert-butyl 3-azetidinylcarbamate hydrochloride,3-n-boc-amino-azetidine-hcl PubChem CID: 22101213 IUPAC Name: tert-butyl N-(azetidin-3-yl)carbamate;hydrochloride SMILES: CC(C)(C)OC(=O)NC1CNC1.Cl