missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)azetidine hydrochloride, 97%, Thermo Scientific™

Catalog No. AAH59695ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59695ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH59695ME Supplier Thermo Scientific Chemicals Supplier No. H59695ME

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 217806-26-3
Molecular Formula C8H17ClN2O2
Molecular Weight (g/mol) 208.686
MDL Number MFCD09839423
InChI Key VLOLOLUAVFGUTD-UHFFFAOYSA-N
Synonym 3-boc-aminoazetidine hydrochloride, tert-butyl azetidin-3-ylcarbamate hydrochloride, 3-n-boc-aminoazetidine hcl, tert-butyl azetidin-3-yl carbamate hydrochloride, tert-butyl n-azetidin-3-yl carbamate hydrochloride, 3-boc-amino azetidine hydrochloride, 3-n-boc-aminoazetidine hydrochloride, 3-boc-amino-azetidine hcl, tert-butyl 3-azetidinylcarbamate hydrochloride, 3-n-boc-amino-azetidine-hcl
PubChem CID 22101213
IUPAC Name tert-butyl N-(azetidin-3-yl)carbamate;hydrochloride
SMILES CC(C)(C)OC(=O)NC1CNC1.Cl

Specifications

Boiling Point 303°C
Quantity 500 mg
Flash Point 137°C (279°F)
Formula Weight 208.69
Percent Purity 97%
Chemical Name or Material 3-(Boc-amino)azetidine hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures