missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Benzylthiazolium Bromide 98.0+%, TCI America™
Supplier: TCI America B193925G
Specifications
| 3-Benzylthiazolium Bromide | |
| 158°C | |
| C10H10BrNS | |
| 25 g | |
| C1=CC=C(C=C1)C[N+]2=CSC=C2.[Br-] | |
| 256.161 | |
| 256.16 | |
| Crystalline Powder |
| 75066-50-1 | |
| White-Yellow | |
| MFCD01631293 | |
| BVVXTLWKNXVYDS-UHFFFAOYSA-M | |
| 3-benzyl-1,3-thiazol-3-ium;bromide | |
| 11608558 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 75066-50-1 | |
| 256.161 | |
| BVVXTLWKNXVYDS-UHFFFAOYSA-M | |
| 3-benzyl-1,3-thiazol-3-ium;bromide |
| C10H10BrNS | |
| MFCD01631293 | |
| 11608558 | |
| C1=CC=C(C=C1)C[N+]2=CSC=C2.[Br-] |
Safety and Handling
TSCA : No