missing translation for 'onlineSavingsMsg'
Learn More

3-Benzylthiazolium Bromide 98.0+%, TCI America™

Catalog No. p-7119368
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19395G 5 g
B193925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19395G Supplier TCI America Supplier No. B19395G
Only null left

Chemical Identifiers

CAS 75066-50-1
Molecular Formula C10H10BrNS
Molecular Weight (g/mol) 256.161
MDL Number MFCD01631293
InChI Key BVVXTLWKNXVYDS-UHFFFAOYSA-M
PubChem CID 11608558
IUPAC Name 3-benzyl-1,3-thiazol-3-ium;bromide
SMILES C1=CC=C(C=C1)C[N+]2=CSC=C2.[Br-]

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 5 g
Formula Weight 256.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzylthiazolium Bromide