missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxyphenyl isothiocyanate, 97%

Catalog No. p-7047443
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19242MD 250 mg
AAL1924203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19242MD Supplier Thermo Scientific Chemicals Supplier No. L19242MD
Only null left

CAS: 206559-36-6 | C14H11NOS | 241.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 206559-36-6
Molecular Formula C14H11NOS
Molecular Weight (g/mol) 241.31
MDL Number MFCD00060374
InChI Key QJLFNDXMZQJMDL-UHFFFAOYSA-N
Synonym 3-benzyloxyphenyl isothiocyanate, 3-benzyloxyphenylisothiocyanate, 1-benzyloxy-3-isothiocyanatobenzene, acmc-20an9f, 3-phenylmethoxy benzenisothiocyanate, 1-benzyloxy-3-isothiocyanatobenzene #, 1-isothiocyanato-3-phenylmethoxy-benzene, benzene, 1-isothiocyanato-3-phenylmethoxy, benzene, 1-isothiocyanato-3-phenylmethoxy-9ci
PubChem CID 561446
IUPAC Name 1-isothiocyanato-3-phenylmethoxybenzene
SMILES S=C=NC1=CC(OCC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 49°C to 51°C
Quantity 250 mg
UN Number UN2923
Sensitivity Moisture sensitive
Formula Weight 241.31
Percent Purity 97%
Chemical Name or Material 3-Benzyloxyphenyl isothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only