missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzyloxy)phenol 95.0+%, TCI America™

Catalog No. p-7119461
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20705G 5 g
B207025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20705G Supplier TCI America Supplier No. B20705G

Chemical Identifiers

CAS 3769-41-3
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00134682
InChI Key FOTVZLOJAIEAOY-UHFFFAOYSA-N
Synonym Resorcinol Monobenzyl Ether
PubChem CID 138048
IUPAC Name 3-phenylmethoxyphenol
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)O

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 200°C
Quantity 5 g
Formula Weight 200.24
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Benzyloxy)phenol