accessibility menu, dialog, popup

UserName

OHO

CAS RN 3769-41-3

IUPAC Name: 3-(benzyloxy)phenol
Synonyms: 3-(benzyloxy)phenol
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: FOTVZLOJAIEAOY-UHFFFAOYSA-N
SMILES: OC1=CC=CC(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results