missing translation for 'onlineSavingsMsg'
Learn More

3-Benzylbenzeneboronic acid pinacol ester, 97%

Catalog No. AAH2868203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2868203 1 g
AAH2868206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2868203 Supplier Thermo Scientific Chemicals Supplier No. H2868203

CAS: 1073355-05-1 | C19H23BO2 | 294.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073355-05-1
Molecular Formula C19H23BO2
Molecular Weight (g/mol) 294.201
MDL Number MFCD09266179
InChI Key VNZSTZMLJUPNQM-UHFFFAOYSA-N
Synonym 2-3-benzylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-benzylphenylboronic acid pinacol ester, 3-benzylbenzeneboronic acid pinacol ester, amtb038
PubChem CID 46739037
IUPAC Name 2-(3-benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CC3=CC=CC=C3

Specifications

Melting Point 68°C to 69°C
Quantity 1 g
Formula Weight 294.2
Percent Purity 97%
Chemical Name or Material 3-Benzylbenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only