missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzylamino)propionitrile, 97+%

Catalog No. p-7051778
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4194914 25 g
AA4194922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4194914 Supplier Thermo Scientific Chemicals Supplier No. 04194914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 706-03-6 | C10H13N2 | 161.23 g/mol

3-(Benzyl amino)propionitrile was used as starting reagent in the synthesis of (3R,4S)-1-benzyl-4-phenylpyrrolidine-3-carboxylic acid, 3-[benzyl-(3,5-dimethyl-pyrazol-1-ylmethyl)-amino]-propionitrile. It undergoes aza-type Michael reaction with 1-hydroxymethyl-5-methyl-1H-pyrazole-3-carboxylic acid ethyl ester under mild conditions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 706-03-6
Molecular Formula C10H13N2
Molecular Weight (g/mol) 161.23
MDL Number MFCD00001955
InChI Key MWTGBAURSCEGSL-UHFFFAOYSA-O
Synonym 3-benzylamino propanenitrile, 3-benzylamino propionitrile, n-benzyl-2-cyanoethylamine, propanenitrile, 3-phenylmethyl amino, a-cyanoethyl benzylamine, beta-cyanoethylbenzylamine, acmc-1bk6i, 3-benzylamino-propionitrile
PubChem CID 69719
IUPAC Name 3-(benzylamino)propanenitrile
SMILES N#CCC[NH2+]CC1=CC=CC=C1

Specifications

Density 1.024 g/mL
Flash Point >110°C (230°F)
Refractive Index 1.5308
Quantity 25 g
UN Number UN3276
Solubility Information Soluble in DMSO.
Formula Weight 160.22
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 3-(Benzylamino)propionitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 211-890-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only