accessibility menu, dialog, popup

UserName

CAS RN 706-03-6

IUPAC Name: benzyl(2-cyanoethyl)azanium
Synonyms: benzyl(2-cyanoethyl)azanium
Molecular Weight (g/mol): 161.23
Molecular Formula: C10H13N2
InChi Key: MWTGBAURSCEGSL-UHFFFAOYSA-O
SMILES: N#CCC[NH2+]CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results