missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylacrylic acid, predominantly trans, 97%

Catalog No. p-7081247
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1114714 25 g
AAA1114722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1114714 Supplier Thermo Scientific Chemicals Supplier No. A1114714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 583-06-2 | C10H8O3 | 176.171 g/mol

2-Butenoic acid, 4-oxo-4-phenyl- could react with morpholine, and obtain the 2-morpholin-4-yl-4-oxo-4-phenyl-butyric acid. This reaction needs the solvent of toluene, ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butenoic acid, 4-oxo-4-phenyl- could react with morpholine, and obtain the 2-morpholin-4-yl-4-oxo-4-phenyl-butyric acid. This reaction needs the solvent of toluene, ethanol.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 583-06-2
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00014015
InChI Key PLPDHGOODMBBGN-VOTSOKGWSA-N
Synonym 3-benzoylacrylic acid, benzoylacrylic acid, 4-oxo-4-phenyl-2-butenoic acid, e-4-oxo-4-phenylbut-2-enoic acid, acrylic acid, 3-benzoyl, 2-butenoic acid, 4-oxo-4-phenyl, 4-oxo-4-phenylbut-2-enoic acid, crotonic acid, 4-oxo-4-phenyl, .beta.-benzoylacrylic acid, trans-3-benzoylacrylic acid
PubChem CID 676116
IUPAC Name (E)-4-oxo-4-phenylbut-2-enoic acid
SMILES C1=CC=C(C=C1)C(=O)C=CC(=O)O

Specifications

Melting Point 93°C to 97°C
Odor Odorless
Quantity 25 g
UN Number UN2811
Beilstein 1817876
Solubility Information Insoluble in water.
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 3-Benzoylacrylic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 209-496-5
RTECSNumber EM9259050
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only