accessibility menu, dialog, popup

UserName

OHOO(E)

CAS RN 583-06-2

IUPAC Name: (2E)-4-oxo-4-phenylbut-2-enoic acid
Synonyms: 3-benzoylacrylic acid (trans)
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: PLPDHGOODMBBGN-VOTSOKGWSA-N
SMILES: OC(=O)\C=C\C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results