missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopropyldimethoxymethylsilane 97.0+%, TCI America™
Supplier: TCI America A262825ML
Specifications
| 3-Aminopropyldimethoxymethylsilane | |
| Yellow | |
| C6H17NO2Si | |
| 25 mL | |
| 3-dimethoxymethylsilyl propylamine, 3-aminopropylmethyldimethoxysilane, 3-dimethoxy methyl silyl propan-1-amine, 3-aminopropyldimethoxymethylsilane, 3-aminopropyl dimethoxymethylsilane, 3-aminopropyl methyl dimethoxy silane, 3-dimethoxy methyl silyl propylamine, gamma-aminopropylmethyldimethoxysilane | |
| CO[Si](C)(CCCN)OC | |
| 163.292 | |
| 163.29 | |
| Liquid |
| 3663-44-3 | |
| 84°C | |
| MFCD00025614 | |
| 3267 | |
| ZYAASQNKCWTPKI-UHFFFAOYSA-N | |
| 3-[dimethoxy(methyl)silyl]propan-1-amine | |
| 77233 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 3663-44-3 | |
| 163.292 | |
| ZYAASQNKCWTPKI-UHFFFAOYSA-N | |
| 77233 | |
| CO[Si](C)(CCCN)OC |
| C6H17NO2Si | |
| MFCD00025614 | |
| 3-dimethoxymethylsilyl propylamine, 3-aminopropylmethyldimethoxysilane, 3-dimethoxy methyl silyl propan-1-amine, 3-aminopropyldimethoxymethylsilane, 3-aminopropyl dimethoxymethylsilane, 3-aminopropyl methyl dimethoxy silane, 3-dimethoxy methyl silyl propylamine, gamma-aminopropylmethyldimethoxysilane | |
| 3-[dimethoxy(methyl)silyl]propan-1-amine |
Safety and Handling
TSCA : No