missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyldimethoxymethylsilane 97.0+%, TCI America™

Catalog No. p-7117998
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A262825ML 25 mL
A2628100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A262825ML Supplier TCI America Supplier No. A262825ML

Chemical Identifiers

CAS 3663-44-3
Molecular Formula C6H17NO2Si
Molecular Weight (g/mol) 163.292
MDL Number MFCD00025614
InChI Key ZYAASQNKCWTPKI-UHFFFAOYSA-N
Synonym 3-dimethoxymethylsilyl propylamine, 3-aminopropylmethyldimethoxysilane, 3-dimethoxy methyl silyl propan-1-amine, 3-aminopropyldimethoxymethylsilane, 3-aminopropyl dimethoxymethylsilane, 3-aminopropyl methyl dimethoxy silane, 3-dimethoxy methyl silyl propylamine, gamma-aminopropylmethyldimethoxysilane
PubChem CID 77233
IUPAC Name 3-[dimethoxy(methyl)silyl]propan-1-amine
SMILES CO[Si](C)(CCCN)OC

Specifications

Color Yellow
Boiling Point 84°C
Quantity 25 mL
UN Number 3267
Formula Weight 163.29
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Aminopropyldimethoxymethylsilane