accessibility menu, dialog, popup

UserName

CH3OSiCH3H2NOH3C

CAS RN 3663-44-3

IUPAC Name: (3-aminopropyl)dimethoxymethylsilane
Synonyms: (3-aminopropyl)dimethoxymethylsilane
Molecular Weight (g/mol): 163.29
Molecular Formula: C6H17NO2Si
InChi Key: ZYAASQNKCWTPKI-UHFFFAOYSA-N
SMILES: CO[Si](C)(CCCN)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results