missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopropionitrile fumarate, 98+%
CAS: 2079-89-2 | (NH2CH2CH2CN)2·HO2CCH=CHCO2H
$101.67 - $372.48
Chemical Identifiers
| CAS | 2079-89-2 |
|---|---|
| Molecular Formula | (NH2CH2CH2CN)2·HO2CCH=CHCO2H |
| MDL Number | MFCD00078321 |
| Synonym | Bis(3-aminopropionitrile) fumarate; 2-Cyanoethylamine hemifumarate |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2544006
|
Thermo Scientific Chemicals
B2544006 |
5 g |
N/A
|
|
|||||
|
AAB2544014
|
Thermo Scientific Chemicals
B2544014 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2079-89-2 | |
| (NH2CH2CH2CN)2·HO2CCH=CHCO2H | |
| MFCD00078321 | |
| Bis(3-aminopropionitrile) fumarate; 2-Cyanoethylamine hemifumarate |
Specifications
| 2079-89-2 | |
| Odorless | |
| MFCD00078321 | |
| 3573532 | |
| 256.26 | |
| 3-Aminopropionitrile fumarate |
| ∼177°C (decomposition) | |
| (NH2CH2CH2CN)2·HO2CCH=CHCO2H | |
| 5 g | |
| Bis(3-aminopropionitrile) fumarate; 2-Cyanoethylamine hemifumarate | |
| ≥98% |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P264b-P270-P280-P301+P312-P308+P313-P330-P501c
H302-H360
EINECSNumber : 218-208-7
RTECSNumber : UG0700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only