accessibility menu, dialog, popup

UserName

NH2NNH2NOHOOHO(E)

CAS RN 2079-89-2

IUPAC Name: (2E)-but-2-enedioic acid; bis(3-aminopropanenitrile)
Synonyms: fumaric acid; bis(β aminopropionitrile)
Molecular Weight (g/mol): 256.26
Molecular Formula: C10H16N4O4
InChi Key: NYMXYZMHOZAPHQ-SEPHDYHBSA-N
SMILES: NCCC#N.NCCC#N.OC(=O)\C=C\C(O)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)