Learn More
3-Aminopropionitrile fumarate, 98+%
CAS: 2079-89-2 | (NH2CH2CH2CN)2·HO2CCH=CHCO2H
Supplier: Thermo Scientific Chemicals B2544006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Aminopropionitrile fumarate | |
| ∼177°C (decomposition) | |
| (NH2CH2CH2CN)2·HO2CCH=CHCO2H | |
| 5 g | |
| Bis(3-aminopropionitrile) fumarate; 2-Cyanoethylamine hemifumarate | |
| ≥98% |
| 2079-89-2 | |
| Odorless | |
| MFCD00078321 | |
| 3573532 | |
| 256.26 |
Chemical Identifiers
| 2079-89-2 | |
| (NH2CH2CH2CN)2·HO2CCH=CHCO2H | |
| MFCD00078321 | |
| Bis(3-aminopropionitrile) fumarate; 2-Cyanoethylamine hemifumarate |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P264b-P270-P280-P301+P312-P308+P313-P330-P501c
H302-H360
EINECSNumber : 218-208-7
RTECSNumber : UG0700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only