missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylacetylene, 98%

CAS: 54060-30-9 | C8H7N | 117.15 g/mol

Supplier:  Thermo Scientific Chemicals L2012414

 View more versions of this product

Catalog No. AAL2012414


Only null left
Explore available promotions

Description

Description

It is used as Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as Pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heavy metal powders.
Specifications

Specifications

3-Aminophenylacetylene
22°C
70°C (1 mmHg)
C8H7N
MFCD00014779
2935417
Insoluble in water.
NC1=CC=CC(=C1)C#C
117.15
117.15
54060-30-9
1.04
>100°C (212°F)
1.617
25 g
3-aminophenylacetylene, benzenamine, 3-ethynyl, m-aminophenylacetylene, 3-ethynylbenzenamine, 3-ethynylphenylamine, 3-ethynylaniline, m-aminophenyl acetylene, 3-aminophenyl acetylene, m-ethynylaniline, 3-ethynyl-aniline
NNKQLUVBPJEUOR-UHFFFAOYSA-N
3-ethynylaniline
104682
98%

Chemical Identifiers

54060-30-9
117.15
NNKQLUVBPJEUOR-UHFFFAOYSA-N
104682
NC1=CC=CC(=C1)C#C
C8H7N
MFCD00014779
3-aminophenylacetylene, benzenamine, 3-ethynyl, m-aminophenylacetylene, 3-ethynylbenzenamine, 3-ethynylphenylamine, 3-ethynylaniline, m-aminophenyl acetylene, 3-aminophenyl acetylene, m-ethynylaniline, 3-ethynyl-aniline
3-ethynylaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 258-944-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only