missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminophenylacetylene, 98%
Description
It is used as Pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as Pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heavy metal powders.
It is used as Pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heavy metal powders.
Specifications
Specifications
| Melting Point | 22°C |
| Density | 1.04 |
| Boiling Point | 70°C (1 mmHg) |
| Flash Point | >100°C (212°F) |
| Refractive Index | 1.617 |
| Quantity | 5 g |
| Beilstein | 2935417 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 117.15 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H312-H315-H319 Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: III |
| EINECSNumber | 258-944-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only