missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenol 98.5+%, TCI America™

Catalog No. p-7116869
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
A038325G 25 g
A0383500G 500 g
2 options

Catalog No. A038325G

Supplier: TCI America A038325G

Only null left

Chemical Identifiers

CAS 591-27-5
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00007786
InChI Key CWLKGDAVCFYWJK-UHFFFAOYSA-N
Synonym m-aminophenol, m-hydroxyaniline, 3-hydroxyaniline, phenol, 3-amino, 1-amino-3-hydroxybenzene, 3-amino-1-hydroxybenzene, fouramine eg, futramine eg, fourrine eg, pelagol eg
PubChem CID 11568
ChEBI CHEBI:28924
IUPAC Name 3-aminophenol
SMILES NC1=CC=CC(O)=C1

Specifications

Melting Point 122°C
Color White-Yellow
Boiling Point 164°C
Quantity 25 g
UN Number 2512
Formula Weight 109.13
Percent Purity ≥98.5% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminophenol
EINECSNumber (3)-0675
RTECSNumber SJ4900000
TSCA Yes