accessibility menu, dialog, popup

UserName

NH2HO

CAS RN 591-27-5

IUPAC Name: 3-aminophenol
Synonyms: m-aminophenol renal EG 3-aminophenol (product) nako teg futramine EG basf ursol EG C.I. oxidation base 7 3-aminophenol (substance) tertral EG CI oxidation base 7
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: CWLKGDAVCFYWJK-UHFFFAOYSA-N
SMILES: NC1=CC=CC(O)=C1
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)