missing translation for 'onlineSavingsMsg'
Learn More

3-(Aminomethyl)benzoic acid hydrochloride, 95%, Thermo Scientific Chemicals

Catalog No. AAH6454603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6454603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6454603 Supplier Thermo Scientific Chemicals Supplier No. H6454603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 876-03-9 | Molecular Formula: C8H10ClNO2 | Molecular Weight: 187.623 g/mol

It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 876-03-9
Molecular Formula C8H10ClNO2
Molecular Weight (g/mol) 187.623
MDL Number MFCD03791117
InChI Key SJCCOASSOPUHEN-UHFFFAOYSA-N
Synonym 3-aminomethyl benzoic acid hydrochloride, 3-aminomethylbenzoic acid hydrochloride, 3-aminomethylbenzoic acid hcl, 3-aminomethyl-benzoic acid hydrochloride, acmc-209tzp, 3-aminomethyl benzoic acid hcl, 3-carboxybenzylamine hydrochloride, 3-aminomethyl-benzoic acid hcl salt, 3-aminomethyl benzoic acid, chloride, 3-aminomethyl benzoicacid hydrochloride
PubChem CID 16218772
IUPAC Name 3-(aminomethyl)benzoic acid;hydrochloride
SMILES C1=CC(=CC(=C1)CN)C(=O)O.Cl

Specifications

Melting Point 242°C to 244°C
Quantity 1 g
Beilstein 3697656
Solubility Information Slightly soluble in water.
Formula Weight 187.62
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(Aminomethyl)benzoic acid hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only