accessibility menu, dialog, popup

UserName

CAS RN 876-03-9

IUPAC Name: 3-(aminomethyl)benzoic acid hydrochloride
Synonyms: 3-(aminomethyl)benzoic acid hydrochloride
Molecular Weight (g/mol): 187.62
Molecular Formula: C8H10ClNO2
InChi Key: SJCCOASSOPUHEN-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results