Learn More
3-Aminoimidazo[1,2-a]pyridine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5872603
Description
It is an intermediate in the production of mutagenic pyrolyzates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an intermediate in the production of mutagenic pyrolyzates.
Solubility
Slightly Soluble in water. (4.9 g/L) (25°C)
Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
Specifications
| 3-Aminoimidazo[1,2-a]pyridine | |
| 114°C to 126°C | |
| MFCD06660421 | |
| imidazo 1,2-a pyridin-3-amine, 3-aminoimidazo 1,2-a pyridine, 3-amino-imidazo 1,2-a pyridine, acmc-20an9p, imidazopyridinamine, imidazo 1,2-a pyridin-3-ylamine, 4-hydroimidazo 1,2-a pyridine-3-ylamine | |
| RTOFMMILZZGGNS-UHFFFAOYSA-N | |
| imidazo[1,2-a]pyridin-3-amine | |
| 432221 | |
| 97% |
| 28036-33-1 | |
| C7H7N3 | |
| 1 g | |
| Slightly Soluble in water. (4.9g/L) (25°C) | |
| C1=CC2=NC=C(N2C=C1)N | |
| 133.154 | |
| 133.15 |
Chemical Identifiers
| 28036-33-1 | |
| 133.154 | |
| RTOFMMILZZGGNS-UHFFFAOYSA-N | |
| 432221 | |
| C1=CC2=NC=C(N2C=C1)N |
| C7H7N3 | |
| MFCD06660421 | |
| imidazo 1,2-a pyridin-3-amine, 3-aminoimidazo 1,2-a pyridine, 3-amino-imidazo 1,2-a pyridine, acmc-20an9p, imidazopyridinamine, imidazo 1,2-a pyridin-3-ylamine, 4-hydroimidazo 1,2-a pyridine-3-ylamine | |
| imidazo[1,2-a]pyridin-3-amine |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only