missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoimidazo[1,2-a]pyridine, 97%, Thermo Scientific Chemicals

Catalog No. p-7039472
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5872603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5872603 Supplier Thermo Scientific Chemicals Supplier No. H5872603

It is an intermediate in the production of mutagenic pyrolyzates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in the production of mutagenic pyrolyzates.

Solubility
Slightly Soluble in water. (4.9 g/L) (25°C)

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.

Chemical Identifiers

CAS 28036-33-1
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD06660421
InChI Key RTOFMMILZZGGNS-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridin-3-amine, 3-aminoimidazo 1,2-a pyridine, 3-amino-imidazo 1,2-a pyridine, acmc-20an9p, imidazopyridinamine, imidazo 1,2-a pyridin-3-ylamine, 4-hydroimidazo 1,2-a pyridine-3-ylamine
PubChem CID 432221
IUPAC Name imidazo[1,2-a]pyridin-3-amine
SMILES C1=CC2=NC=C(N2C=C1)N

Specifications

Melting Point 114°C to 126°C
Quantity 1 g
Solubility Information Slightly Soluble in water. (4.9g/L) (25°C)
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 3-Aminoimidazo[1,2-a]pyridine
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only