accessibility menu, dialog, popup

UserName

CAS RN 28036-33-1

IUPAC Name: imidazo[1,2-a]pyridin-3-amine
Synonyms: imidazo[1,2-a]pyridin-3-amine
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: RTOFMMILZZGGNS-UHFFFAOYSA-N
SMILES: NC1=CN=C2C=CC=CN12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results