missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminobenzylamine 98.0+%, TCI America™
$82.02 - $225.56
Chemical Identifiers
| CAS | 4403-70-7 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.17 |
| MDL Number | MFCD00078355 |
| InChI Key | ZDBWYUOUYNQZBM-UHFFFAOYSA-N |
| Synonym | 3-aminobenzylamine, 3-aminomethyl aniline, m-aminobenzylamine, benzenemethanamine, 3-amino, 3-aminomethyl-aniline, pubchem5802, alpha-amino-m-toluidine, 3-aminomethyl phenylamine, 3-aminophenyl methylamine |
| PubChem CID | 4628831 |
| IUPAC Name | 3-(aminomethyl)aniline |
| SMILES | NCC1=CC(N)=CC=C1 |
Chemical Identifiers
| 4403-70-7 | |
| 122.17 | |
| ZDBWYUOUYNQZBM-UHFFFAOYSA-N | |
| 4628831 | |
| NCC1=CC(N)=CC=C1 |
| C7H10N2 | |
| MFCD00078355 | |
| 3-aminobenzylamine, 3-aminomethyl aniline, m-aminobenzylamine, benzenemethanamine, 3-amino, 3-aminomethyl-aniline, pubchem5802, alpha-amino-m-toluidine, 3-aminomethyl phenylamine, 3-aminophenyl methylamine | |
| 3-(aminomethyl)aniline |
Specifications
| 4403-70-7 | |
| White-Yellow | |
| C7H10N2 | |
| 5 g | |
| 3-aminobenzylamine, 3-aminomethyl aniline, m-aminobenzylamine, benzenemethanamine, 3-amino, 3-aminomethyl-aniline, pubchem5802, alpha-amino-m-toluidine, 3-aminomethyl phenylamine, 3-aminophenyl methylamine | |
| NCC1=CC(N)=CC=C1 | |
| 122.17 | |
| 122.17 | |
| Crystalline Powder |
| 43°C | |
| 134°C | |
| MFCD00078355 | |
| 3259 | |
| ZDBWYUOUYNQZBM-UHFFFAOYSA-N | |
| 3-(aminomethyl)aniline | |
| 4628831 | |
| ≥98.0% (GC,T) | |
| 3-Aminobenzylamine |
Safety and Handling
EINECSNumber : (3)-3897
TSCA : No