accessibility menu, dialog, popup

UserName

CAS RN 4403-70-7

IUPAC Name: 3-(aminomethyl)aniline
Synonyms: 3-(aminomethyl)aniline
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: ZDBWYUOUYNQZBM-UHFFFAOYSA-N
SMILES: NCC1=CC(N)=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)