missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzenethiol 97.0+%, TCI America™

Catalog No. p-7117605
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18525G 5 g
A185225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18525G Supplier TCI America Supplier No. A18525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22948-02-3
Molecular Formula C6H6NS
Molecular Weight (g/mol) 124.18
MDL Number MFCD00007791
InChI Key KFFUEVDMVNIOHA-UHFFFAOYSA-M
Synonym 3-aminothiophenol, benzenethiol, 3-amino, 3-mercaptoaniline, m-amino thiophenol, 3-aminobenzene-1-thiol, 3-amino thiophenol, 3-amino-benzenethiol, pubchem2470, acmc-1co9c
PubChem CID 31577
IUPAC Name (3-aminophenyl)sulfanide
SMILES NC1=CC=CC([S-])=C1

Specifications

Boiling Point 190°C
Quantity 5 g
UN Number 3267
Formula Weight 125.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Aminobenzenethiol