accessibility menu, dialog, popup

UserName

CAS RN 22948-02-3

IUPAC Name: (3-aminophenyl)sulfanide
Synonyms: (3-aminophenyl)sulfanide
Molecular Weight (g/mol): 124.18
Molecular Formula: C6H6NS
InChi Key: KFFUEVDMVNIOHA-UHFFFAOYSA-M
SMILES: NC1=CC=CC([S-])=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)