missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-N-methylbenzylamine 97.0+%, TCI America™

Catalog No. A21585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21585G 5 g
A215825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21585G Supplier TCI America Supplier No. A21585G

Chemical Identifiers

CAS 18759-96-1
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00211072
InChI Key HBLPYXIZPMDWIO-UHFFFAOYSA-N
Synonym 3-(Methylaminomethyl)aniline
PubChem CID 11643936
IUPAC Name 3-[(methylamino)methyl]aniline
SMILES CNCC1=CC(N)=CC=C1

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3259
Formula Weight 136.20
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Amino-N-methylbenzylamine